missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iBind™ Flex Fluorescent Detection (FD) Solution Kit
Detection kit for use with iBind™ Flex Western System
Brand: Invitrogen™ SLF2019
110.20 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
iBind™ Flex Solution is a proprietary formulation compatible with downstream fluorescent detection/chemiluminescent detection. It serves as a combined blocking, washing and antibody diluent solution to process up to one midi blot, two mini blots, or six vertically cut strip blots when used with the iBind™ Flex Western Device and iBind™ Flex Cards.Each kit contains reagents sufficient for 10 midi blots or 20 mini blots
Fluorescent Detection (FD) Solution (Contents)
- iBind Flex FD 5X Buffer
- iBind Flex 100X Additive
- iBind Flex FD 10% SDS
Chemiluminescent Solution (Contents)
- iBind Flex 5X Buffer
- iBind Flex 100X Additive
Specifications
Flex Solution Kit | |
iBind Flex Western Device | |
10 midi blots or 20 mini blots | |
Fluorescence | |
iBind™ |
• iBind Flex FD 5X Buffer • iBind Flex 100X Additive • iBind Flex FD 10% SDS Store at 2–8°C. |
|
iBind™ Flex FD 5X buffer, iBind™ Flex 100X additive, iBind™ Flex FD 10% SDS | |
Approved for shipment at Room Temperature or on Wet Ice | |
PVDF, Nitrocellulose | |
1 kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- iBind Flex Fluorescent Detection (FD) Solution Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : proclin 200, 5-chloro-2-methyl-3(2H)-isothiazolone, 2-methyl-3(2H)-isothiazolone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction