missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Solution Kit

Product Code. p-5013595
Click to view available options
Quantity:
1 kit
4 kits
Product Type:
Flex Solution Kit
Unit Size:
1 set
Pack of 4
This item is not returnable. View return policy

Product Code. 15357248

Brand: Invitrogen™ SLF2020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detection kit for use with iBind™ Flex Western System

iBind™ Flex Solution is a proprietary formulation compatible with downstream fluorescent detection/chemiluminescent detection. It serves as a combined blocking, washing and antibody diluent solution to process up to one midi blot, two mini blots, or six vertically cut strip blots when used with the iBind™ Flex Western Device and iBind™ Flex Cards.

Each kit contains reagents sufficient for 10 midi blots or 20 mini blots

Fluorescent Detection (FD) Solution (Contents)

  • iBind Flex FD 5X Buffer
  • iBind Flex 100X Additive
  • iBind Flex FD 10% SDS

Chemiluminescent Solution (Contents)

  • iBind Flex 5X Buffer
  • iBind Flex 100X Additive

Store at 2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Product Type Flex Solution Kit
Content And Storage • iBind Flex 5X Buffer
• iBind Flex 100X Additive

Store at 2–8°C.
For Use With (Equipment) iBind Flex Western Device
Kit Contents iBind™ Flex 5X buffer, iBind™ Flex 100X additive
No. of Reactions 10 midi blots or 20 mini blots
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Detection Method Colorimetric, Chromogenic, Chemiluminescence
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind
Quantity 1 kit
compliance-icons
Product Identifier
  • Flex Solution Kit
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-3(2H)-isothiazolone

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.