Learn More
Invitrogen™ iBind™ Solution Kit
Beschreibung
Beinhaltet
Kit contains sufficient solution for 40 mini-membrane reactions.
Benötigt
iBind Western Device
The automated iBind Western Device performs every step in the Western processing workflow from blocking to washes to antibody incubations. The iBind solutions act as the blocking, washing, and dilution solutions in the iBind immunodetection process and have been optimized for ideal viscosity. Each solution is released from the iBind device wells to the iBind Card through sequential lateral flow (SLF); the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.
- Optimized solutions with ideal viscosity for blocking, washing, and antibody dilution
- Long shelf life—store iBind Solutions Kit for 12 months at 4°C
- Better antigen-antibody interaction—solutions flow between the iBind Card and membrane for maximum interaction between protein and antibody
- Brings automation and simplicity to the Western protocol
Proteins, Expression, Isolation and Analysis, Western Blotting
Spezifikation
Spezifikation
| Content And Storage | • iBind 5X buffer (240 mL) • iBind 100X additive (8 x 1.5 mL) Store at 2–8°C. |
| Detection Method | Colorimetric, Chromogenic, Chemiluminescence |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | iBind Western Device |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | iBind |
| Throughput | 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card |
| Type | Solution Kit |
| Includes | iBind 5X buffer (240 mL); iBind 100X additive (8 x 1.5 mL) |
| No. of Reactions | 10 mini membrane reactions |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.