Learn More
Invitrogen™ iBind™ Western Device
An automated Western-processing device that performs every step from blocking to washes to antibody incubations via sequential lateral flow
Brand: Invitrogen™ SLF1000
796.48 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Description
Sequential lateral flow (SLF) allows timely release and flow of solutions and antibodies to the membrane without need of an external power source. Each solution is released from the iBind device wells to an iBind Card via SLF; the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.
- Simple, automated solution
- No external power source
- Superior reproducibility over traditional manual methods
- Better antigen antibody interaction—solutions flow between the iBind Card and membrane for maximum interaction between protein and antibody
- Minimizes hands-on time
- Highly reproducible and compatible with all Western protocols
- Set up time is fast—ready for the detection step within 2.5 hr.
- Carry out detection right away or wait until the next day without affecting performance
- Works exclusively with iBind Cards and the iBind Solution Kit (available separately) to process one mini-membrane for immunodetection
- Provides automated convenience and superior reproducibility over traditional manual methods
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Store at room temperature. | |
Fluorescence, Chemiluminescence, Chromogenic | |
Nitrocellulose, PVDF | |
∼2.5 hr. | |
Western Device |
1 Mini Blot | |
Western Blot | |
iBind | |
1 Mini Blot | |
1 device |
Safety and Handling
- iBind Western Device
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P314-Get medical advice/attention if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formamide
For Research Use Only. Not for use in diagnostic procedures.