missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Ibotenic acid monohydrate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 141.0°C to 147.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 5 mg |
Merck Index | 14, 4877 |
Solubility Information | Solubility in water: soluble in water. |
Formula Weight | 176.1 |
Percent Purity | 95% |
Physical Form | Powder or Solid |
Show More |
Safety and Handling
Product Identifier
- (±)-Ibotenic acid monohydrate
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'