Learn More
Invitrogen™ iBright™ System Contrast Tray
This iBright System Contrast Tray is an iBright imaging system accessory designed to improve the imaging of visible protein gels.
Brand: Invitrogen™ A56597
Additional Details : Weight : 0.35000kg
Description
- This iBright System Contrast Tray is an iBright imaging system accessory designed to improve the imaging of visible protein gels
- The dual layer tray is designed to create optimal lighting conditions when used with the green LED transilluminator on the iBright imaging system, improving the contrast and lowering the overall background to capture high-quality visible-stain images
- Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
- Provides a contrasting background when imaging protein gels stained with colorimetric stains, such as Coomassie blue and silver stains
- This tray also improves conditions when samples contain dyes that auto-fluoresce under green LED light
- Recommended for transillumination of crystal violet-stained colony plates
- A replacement for the White Transilluminator Screen (Cat. No. A33828)
Specifications
Room Temperature | |
iBright | |
Contrast Tray |
1 Tray | |
Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging |
For Research Use Only. Not for use in diagnostic procedures.