Learn More
Invitrogen™ iBright™ System Tray Adapter for E-Gel™ Agarose Gels
Imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.
£130.00
Specifications
Quantity | 1 Adaptor |
---|---|
Shipping Condition | Room Temperature |
Product Line | iBright |
For Use With (Application) | Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
17806521
|
Invitrogen™
A56600 |
E-Gel Tray Adapter, 11-/22-well |
£130.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
17896511
|
Invitrogen™
A56599 |
E-Gel Tray Adapter, 48-/96-well |
£130.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
Description
Invitrogen iBright Tray Adapters for E-Gel Agarose Gels are imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.
Features of Invitrogen iBright Tray Adapters for E-Gel Agarose Gels include:
• The bottom component centers the E-Gel agarose gels on the iBright transilluminator box facilitating optimized zoom
• The top component covers labels and bar codes on the E-Gel cassette to reduce stray light during image capture and improve sample signal optimization
• Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
• Compatible with 11- to 96-well E-Gel agarose (Cat. No. A56600 for 11-/22-well, Cat. No. A56599 for 48-/96-well)
Specifications
1 Adaptor | |
Room Temperature | |
iBright | |
Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging |