missing translation for 'onlineSavingsMsg'
Learn More

Ibrutinib-biotin, MedChemExpress

Product Code. p-200096776
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449526

Brand: Medchem Express HY100342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.

Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.

Specifications

Chemical Name or Material Ibrutinib-biotin
CAS 1599432-18-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C56H80N12O9S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (45.56 mM; Need ultrasonic)
SMILES O=C(CCCC(NCCCOCCOCCOCCCNC(CCCC[C@@H]1SC[C@H](N2)[C@@H]1NC2=O)=O)=O)NCCCCN(C)C/C=C/C(N3CCC[C@@H](N4N=C(C5=CC=C(OC6=CC=CC=C6)C=C5)C7=C(N)N=CN=C74)C3)=O
Molecular Weight (g/mol) 1097.37
Formula Weight 1097.37
Percent Purity 98.04%
Grade Research
Physical Form Oil
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.