missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ibrutinib
Potent and selective BTK inhibitor (IC50= 0.5 nM)
£147.00 - £583.00
Specifications
Chemical Name or Material | 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2-propen-1-one |
---|---|
CAS | 936563-96-1 |
Synonym | Agammaglobulinaemia tyrosine kinase, AGMX1, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase |
Molecular Formula | C25H24N6O2 |
Formula Weight | 440.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757926
|
Tocris Bioscience™
6813/10 |
10 mg |
£147.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18716506
|
Tocris Bioscience™
6813/50 |
50 mg |
£583.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Selective for BTK against a screening panel of kinase enzymes
- Inhibits autophosphorylation of BTK, phosphorylation of PLCγ, and phosphorylation of ERK (IC50 of 11 nM, 29 nM and 13 nM, respectively)
- Induces cytotoxicity of chronic lymphocytic leukemia (CLL) cells
- Reduces clinical arthritis scores in a mouse model.
Specifications
1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2-propen-1-one | |
Agammaglobulinaemia tyrosine kinase, AGMX1, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase | |
440.5 |
936563-96-1 | |
C25H24N6O2 | |
98% |
Safety and Handling
Recommended Storage : Store at -20°C