missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IC Fixation Buffer
IC Fixation Buffer
Brand: Invitrogen™ FB001
Code : 50
Additional Details : Weight : 0.10000kg
Description
Intracellular fixation buffer for use in flow cytomerty, particularly for fixation in intracellular staining procedures. Composition is paraformaldehyde in phosphate buffered saline, pH 7.3. These Make-to-Order catalog items have a 2 week lead time.Specifications
IC Fixation buffer | |
IC Fixation Buffer, 100mL, Colorless/Clear, Liquid, 7.3 pH, 2 to 8°C Storage, For Use with Cellular Imaging, Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation | |
7.3 | |
100 mL | |
1 Each |
Colorless | |
Flow Cytometer | |
Liquid | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.