missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD IC-XT Pack Sabouraud Dextrose Agar with Lecithin and Polysorbate 80
BD IC-XT Pack Sabouraud Dextrose Agar with Lecithin and Polysorbate 80 - Ready-for-use prepared bottled media help detect microorganisms from pharmaceutical, food, environmental or water testing samples.
Brand: BD 257662
Additional Details : Weight : 4.27700kg
Description
- X100 Sabouraud Agar with Lecithin and Polysorbate 80
- IC-XT Pack
- Custom-made
Specifications
Liquid | |
100 | |
90 mm |
Sabouraud Dextrose Agar | |
Plates | |
4.28 kg |