missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Ice Cream Sampler

Constructed of stainless steel and comes complete with a test tube for putting the sample into. Sampling Systems™ Ice Cream Sampler is designed to be easily dismantled for cleaning and is ideal for sampling products like ice cream and soft waxes.

Brand:  Sampling Systems™ 6006A-100

Additional Details : Weight : 0.04500kg

Product Code. 15899371

  • £54.50 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

Description

Description

  • 316L Stainless Steel construction
  • Clear polystyrene test tube
  • Designed to be easily dismantled for cleaning
Specifications

Specifications

Stainless Steel
Sampler and one test tube
25 mm
Ice Cream Sampler
30 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers