missing translation for 'onlineSavingsMsg'
Learn More

ICI 199,441 hydrochloride, Tocris Bioscience™

Product Code. 18085652
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent κ agonist

Highly potent κ agonist, 146-fold more active than U-50488 (Cat. No. 0495) in vitro.

Chemical Identifiers

CAS 115199-84-3
Molecular Formula C21H25Cl3N2O
Molecular Weight (g/mol) 427.794
InChI Key VFLWVWZSDBTGQJ-VEIFNGETSA-N
Synonym ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s
PubChem CID 3082717
IUPAC Name 2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride
SMILES CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl

Specifications

Quantity 50 mg
Formula Weight 427.8
Percent Purity >99%
Chemical Name or Material ICI 199, 441 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'