missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ICK Recombinant Human Protein

ICK or intestinal cell kinase belongs to a very extensive family of proteins that share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. Full-length recombinant human ICK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

Brand:  Invitrogen™ PV6359

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15448993

  • £1460.00 / 100µg
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

ICK encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. ICK localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. ICK purified from transfected human embryonic kidney cells undergoes autophosphorylation and phosphorylates a test protein. ICK plays a key role in development of central nervous, skeletal, and endocrine systems.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
95.1kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
ECO, MRK, KIAA0936, LCK2, MGC46090
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers