missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ CO2 Incubator ICO
Ideal for Incubation with CO2, Biosyntesis and Cell Incubation in the Laboratory.
Brand: Memmert™ ICO105
4397.26 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Sterile, safe and reliable – these three words describe the Memmert CO2 incubator ICO for industrial use.
- Model sizes: 50, 105, 150, 240
- Temperature adjustment range: +18 °C to +50 °C
- CO2 adjustment range: 0 to 20 %
- Humidity adjustment ranges: 40 to 97 % rh (with option K7)
- O2 adjustment ranges: 1 to 20 % (with option T6)
- 1 model variant: TwinDISPLAY (intuitive control)
- AtmoCONTROL software on a USB stick for programming, managing and transferring programmes via Ethernet interface or USB port
- Perfect distribution of temperature, humidity and CO2
- Long-time stable and repetitively accurate
- The interior chamber, including all installations and sensors, can be sterilised at +180 °C in a 60-minute programme
- Visual and acoustic alarm as well as alarm notification to mobile phones if the individually adjustable temperature, CO2, O2, or humidity ranges are exceeded
- Battery-buffered ControlCOCKPIT: The operating display, logging and CO2 control are fully functional even during a power failure
- Fail-safe, FDA-compliant logging of temperature, CO2, O2 humidity, open door time data
- Adjustable parameters: temperature (Celsius or Fahrenheit), CO2, programme time, time zones, summertime/wintertime
Active humidity control ensures short recovery times after the door has been opened and minimises vaporisation in the interior. Together with the heating of the interior from all six sides and the heated inner glass door, it offers maximum protection for cell and tissue cultures and avoids the dangerous formation of condensation. The turbulence-free chamber ventilation ensures a constant and uniform atmosphere.
Specifications
Optic and acoustic | |
TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays | |
Stainless Steel | |
IR Sensor | |
2 | |
40 to 97% | |
CO2 Incubator | |
2 high-definition TFT-colour displays (TwinDISPLAY) | |
Digital backwards counter with target time setting, adjustable from 1 minute to 99 days | |
591 x 719 x 846 mm | |
75 kg | |
115/230 V |
15 kg | |
107 L | |
0 to 20% | |
Textured Stainless Steel, Rear Zinc-plated Steel | |
1 to 20% | |
18°C to 50°C | |
60 min. | |
Perforated stainless steel | |
400 x 560 x 480 mm | |
100 kg | |
115/230 V, 50/60 Hz | |
50/60 Hz |