Learn More
Calibration Standard 3 For ICP/MS, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CLMS-3
Code : 04
Additional Details : Weight : 0.12500kg Transport : UN number : 3264 Chem class : 8 Pack group : II
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Au | 10μg/mL |
Hf | 10μg/mL |
Ir | 10μg/mL |
Pd | 10μg/mL |
Pt | 10μg/mL |
Rh | 10μg/mL |
Ru | 10μg/mL |
Sb | 10μg/mL |
Sn | 10μg/mL |
Te | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Multi-Element Solution 3 | |
125 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Each component at 10μg/mL in 10% HCl/1% HNO3: Antimony Gold Hafnium Iridium Palladium Platinum Rhodium Ruthenium Tellurium Tin |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Trace ICP/ICP-MS | |
10% HCl/1% HNO3 |
Product intended for laboratory use only