missing translation for 'onlineSavingsMsg'
Learn More

IEICO, Thermo Scientific Chemicals

£471.35 - £1602.98

Chemical Identifiers

CAS 2055812-53-6
Molecular Formula C114H118N4O4S4
Molecular Weight (g/mol) 1736.46
InChI Key MLLSAVAQBWNXIZ-UHFFFAOYNA-N
Synonym 2,2 '-((2 Z,2 'Z)-((5,5 '-bis(4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(4-((2- ethylhexyl)oxy)thiophene-5,2-diyl))bis(methanylylidene))bis- (3-oxo-2,3-dihydro-1 H-indene-2,1-diylidene))dimalononitrile
IUPAC Name 2-[2-({5-[15-(5-{[1-(dicyanomethylidene)-3-oxo-2,3-dihydro-1H-inden-2-ylidene]methyl}-3-[(2-ethylhexyl)oxy]thiophen-2-yl)-9,9,18,18-tetrakis(4-hexylphenyl)-5,14-dithiapentacyclo[10.6.0.0³,¹⁰.0⁴,⁸.0¹³,¹⁷]octadeca-1(12),2,4(8),6,10,13(17),15-heptaen-6-yl]-4-[(2-ethylhexyl)oxy]thiophen-2-yl}methylidene)-3-oxo-2,3-dihydro-1H-inden-1-ylidene]propanedinitrile
SMILES CCCCCCC1=CC=C(C=C1)C1(C2=C(SC(=C2)C2=C(OCC(CC)CCCC)C=C(S2)C=C2C(=O)C3=CC=CC=C3C2=C(C#N)C#N)C2=CC3=C(C=C12)C1=C(C=C(S1)C1=C(OCC(CC)CCCC)C=C(S1)C=C1C(=O)C2=CC=CC=C2C1=C(C#N)C#N)C3(C1=CC=C(CCCCCC)C=C1)C1=CC=C(CCCCCC)C=C1)C1=CC=C(CCCCCC)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17146564
SDS View Documents
Thermo Scientific Alfa Aesar
H66656.MC
100 mg
£471.35
100mg
Please to purchase this item. Need a web account? Register with us today!
17156564
SDS View Documents
Thermo Scientific Alfa Aesar
H66656.MD
250 mg
£896.46
250mg
Please to purchase this item. Need a web account? Register with us today!
17166564
SDS View Documents
Thermo Scientific Alfa Aesar
H66656.ME
500 mg
£1602.98
500mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IEICO, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.