Learn More
Ifosfamide, Thermo Scientific Chemicals
£104.52 - £377.00
Chemical Identifiers
CAS | 3778-73-2 |
---|---|
Molecular Formula | C7H15Cl2N2O2P |
Molecular Weight (g/mol) | 261.09 |
InChI Key | HOMGKSMUEGBAAB-UHFFFAOYSA-N |
Synonym | ifosfamide, isophosphamide, iphosphamide, isofosfamide, ifosfamid, mitoxana, naxamide, iphosphamid, isoendoxan, ifex |
PubChem CID | 3690 |
ChEBI | CHEBI:5864 |
IUPAC Name | N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine |
SMILES | C1CN(P(=O)(OC1)NCCCl)CCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15628735
|
Thermo Scientific Acros
461350010 |
1 g |
£104.52
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15638735
|
Thermo Scientific Acros
461350050 |
5 g |
£377.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3778-73-2 | |
261.09 | |
ifosfamide, isophosphamide, iphosphamide, isofosfamide, ifosfamid, mitoxana, naxamide, iphosphamid, isoendoxan, ifex | |
CHEBI:5864 | |
C1CN(P(=O)(OC1)NCCCl)CCCl |
C7H15Cl2N2O2P | |
HOMGKSMUEGBAAB-UHFFFAOYSA-N | |
3690 | |
N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine |
Specifications
3778-73-2 | |
Authentic | |
ifosfamide, isophosphamide, iphosphamide, isofosfamide, ifosfamid, mitoxana, naxamide, iphosphamid, isoendoxan, ifex | |
C1CN(P(=O)(OC1)NCCCl)CCCl | |
261.09 | |
CHEBI:5864 | |
≥97.5% (ELSD) (HPLC) |
39.0°C to 42.0°C | |
C7H15Cl2N2O2P | |
HOMGKSMUEGBAAB-UHFFFAOYSA-N | |
N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine | |
3690 | |
261.09 | |
Ifosfamide |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-237-3
RUO – Research Use Only