missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Mouse anti-Human, Clone: 3A5A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60099-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes the Chain C region of human Ig alpha-1 (IGHA1).
Ig alpha is the major immunoglobulin class in body secretions. The protein may serve both to defend against local infection and to prevent access of foreign antigens to the general immunologic system.Specifications
Human IgA | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01876 | |
IGHA1 | |
Human IgA Fusion Protein Ag7417 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3A5A5 | |
Unconjugated | |
IGHA1 | |
IgA1, IGHA1 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
3493 | |
-20°C | |
Liquid |