Learn More
Invitrogen™ Mouse IgA ELISA Kit
ELISA
£578.00 - £4605.00
Specifications
Assay Range | 2.05 to 500 ng/mL |
---|---|
Assay Sensitivity | 1.5 ng/mL |
Calibration Range | 1.5 ng/mL to 500 ng/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | HRP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425283
|
Invitrogen™
EMIGA |
96 Tests |
£578.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17125739
|
Invitrogen™
EMIGAX5 |
5 x 96 Tests |
£2450.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18527824
|
Invitrogen™
EMIGAX10 |
10 x 96 Tests |
£4605.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Mouse Immunoglobulin A (Ms IgA) ELISA quantitates Ms IgA in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IgA. Principle of the method The Mouse IgA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Mouse IgA is an immunoglobulin A antibody that is produced by B cells in response to an antigen. It is a crucial component of the mucosal immune system, which protects the body from pathogens that enter through mucosal surfaces such as the respiratory, gastrointestinal, and urogenital tracts. Mouse IgA is composed of two heavy chains and two light chains, and it is the most abundant immunoglobulin in mucosal secretions. It plays a vital role in neutralizing pathogens and preventing their attachment to mucosal surfaces. Mouse IgA is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Specifications
2.05 to 500 ng/mL | |
1.5 ng/mL to 500 ng/mL | |
HRP | |
Other Proteins | |
This ELISA pair antibody detects mouse IgA. Other species not determined. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
238447 | |
<10 | |
HRP | |
Immunology | |
2°C to 8°C | |
1 hrs 20 mins |
1.5 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA | |
Plasma, Serum, Supernatant | |
Mouse | |
Colorimetric Microplate Reader | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
Plasma 2 μL, Serum 2 μL, Supernatant 100 μL | |
Mouse IgA | |
4 hrs 45 mins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.