missing translation for 'onlineSavingsMsg'
Learn More

IGEPAL∣r CA-630

Product Code. 15443719
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 15443719

Brand: Thermo Scientific Alfa Aesar J61055.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Dieses Produkt enthält Substanzen aus dem Anhang XIV der Verordnung REACH (EG) Nr. 1907/2006. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.
Regulatorischer Hinweis: Dieses Produkt ist ein isoliertes Zwischenprodukt, das gemäß der Verordnung (EG) Nr. 1907/2006 (REACH) transportiert wird. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.

IGEPAL∣r CA-630, CAS # 9002-93-1, is a nonionic and non-denaturing detergent used mainly for the solubilization, isolation, and purification of membrane protein complexes.

Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins, IGEPAL™ CA-630 is used as a detergent in paper industry, hand cleaners, metal cleaners. It is suitable for solubilization, isolation and purification of membrane protein complexes and in two-dimensional gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • IGEPAL|r CA-630 is a nonionic and non-denaturing detergent similar to Nonidet P-40

Application

  • IGEPAL|r CA-630 is mainly used for the solubilization, isolation, and purification of membrane protein complexes and in two-dimensional gel electrophoresis
  • IGEPAL|r CA-630 can be used in the composition of homogenizing buffers during western blotting technique in mouse muscle homogenates
  • It has been used in X-gal staining to study the lacZ expression
  • IGEPAL|r CA-630 is also a component of the cell lysis buffer in order to prepare RT-qPCR-ready cell lysates
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Octylphenyl-polyethylene glycol
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Spezifikation

Color Colorless
Physical Form Liquid
Density 1.060
Quantity 100 mL
Solubility Information Miscible with water.
Chemical Name or Material IGEPAL™ CA-630
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • IGEPAL CA-630
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contains : IGEPAL CA-630

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt