missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Donkey anti-Goat, Rhodamine, Secondary Antibody, Proteintech
Donkey anti-Goat Affinipure Secondary Antibody
Brand: Proteintech SA00007-3-500UL
This item is not returnable.
View return policy
Specifications
IgG(H+L) | |
Rhodamine | |
Donkey | |
500 μL | |
Goat | |
Liquid |
Immunofluorescence, Flow Cytometry | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |