missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Donkey anti-Mouse, CoraLite488, Secondary Antibody, Proteintech
Donkey anti-Mouse Affinipure Secondary Antibody
Brand: Proteintech SA00013-5-200UL
38.00 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Specifications
IgG(H+L) | |
CoraLite 488 | |
Donkey | |
200 μL | |
Mouse | |
Liquid |
Immunofluorescence, Flow Cytometry | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |