missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Goat anti-Rabbit, AMCA, Secondary Antibody, Proteintech
Goat anti-Rabbit Affinipure Secondary Antibody
£40.00 - £84.00
Specifications
Antigen | IgG(H+L) |
---|---|
Applications | Immunofluorescence, Flow Cytometry |
Conjugate | AMCA |
Form | Liquid |
Host Species | Goat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17620377
|
Proteintech
SA00010-2-200UL |
200 μL |
£40.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17630377
|
Proteintech
SA00010-2-500UL |
500 μL |
£84.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
IgG(H+L) | |
AMCA | |
Goat | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
Immunofluorescence, Flow Cytometry | |
Liquid | |
Rabbit | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |