missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Goat anti-Rat, AP, Secondary Antibody, Proteintech
Goat anti-Rat Affinipure Secondary Antibody
Brand: Proteintech SA00002-9-1ML
Additional Details : Weight : 0.00010kg
Specifications
IgG(H+L) | |
Alkaline Phosphatase (AP) | |
Goat | |
1 mL | |
Rat | |
Liquid |
ELISA, Western Blot | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |