Learn More
IgG(H+L) Rabbit anti-Goat, Biotin-SP, Secondary Antibody, Proteintech
Rabbit anti-Goat Affinipure Secondary Antibody
Brand:  Proteintech SA00004-4-1ML
Product Code. 17640347
  • £149.00 / 1mL
Explore more special offers
Specifications
IgG(H+L)
Biotin-SP
Rabbit
1 mL
Goat
Liquid
ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.