missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Rabbit anti-Sheep, HRP, Secondary Antibody, Proteintech

Rabbit anti-Sheep Affinipure Secondary Antibody

£40.00 - £149.00

Specifications

Antigen IgG(H+L)
Applications ELISA, Western Blot
Conjugate HRP
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17670297
View Documents
Proteintech
SA00001-16-200UL
200 μL
£40.00
200µL
Please to purchase this item. Need a web account? Register with us today!
17660297
View Documents
Proteintech
SA00001-16-1ML
1 mL
£149.00
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

IgG(H+L)
HRP
Rabbit
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
ELISA, Western Blot
Liquid
Sheep
Secondary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications