missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2a/2b Rat anti-Mouse, Oligo, Clone: R2-40, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940442
This item is not returnable.
View return policy
Description
IgG2a/2b Monoclonal antibody specifically detects IgG2a/2b in Mouse samples. It is validated for Single Cell 3' SequencingSpecifications
IgG2a/2b | |
Monoclonal | |
Oligo | |
IgG2a (Ighg2a; Igh-1); IgG2b (Ighg2b; Igh-3; gamma2b) | |
Pooled Mouse Ig | |
RUO | |
Mouse | |
IgG1 κ |
Single Cell 3' Sequencing | |
Clone R2-40 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |