missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2a Isotype Control, BUV496, BD Biosciences™
Monoclonal Isotype Control
Brand: BD Biosciences 612957
Additional Details : Weight : 0.00030kg
Description
The G155-178 clone has an unknown specificity. Trinitrophenol (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to immunoglobulin Fc receptors. The immunoglobulin secreted by the G155-178 hybridoma was selected as a mouse IgG2a, κ isotype control following screening for low background binding on a variety of mouse and human tissues. It is validated for Flow Cytometry, ControlSpecifications
IgG2a | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
IgG2a kappa Isotype Control | |
50 μg | |
Isotype Control (Primary) | |
IgG2a κ |
Flow Cytometry | |
G155-178 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
TNP-keyhole limpet hemocyanin | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimum conditions, and unconjugated antibody and free BD Horizon BUV496 were removed. |