missing translation for 'onlineSavingsMsg'
Learn More

IgG2a Mouse, CoraLite 488, Clone: 11A1B2, Isotype Control, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-66360-2-100UL

Additional Details : Weight : 0.00010kg

Product Code. 17778054

  • £90.30 / 100µL
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

The Mouse IgG2a isotype control antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of Mouse IgG2a isotype control. This antibody recognizes megathura crenulata antigen. The Mouse IgG2a isotype control antibody has been validated for the following applications: FC analysis. This antibody is coralite 488.

Specifications

Specifications

IgG2a
11A1B2
CoraLite 488
Mouse
Protein A Purification
Primary
Store at -20°C
IgG2a
Flow Cytometry
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3
Recombinant Protein
100 μL
M. crenulata
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers