missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGLL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549022
Additional Details : Weight : 0.01000kg
Description
This gene encodes one of the immunoglobulin lambda-like polypeptides. It is located within the immunoglobulin lambda locus but it does not require somatic rearrangement for expression. The first exon of this gene is unrelated to immunoglobulin variable genes; the second and third exons are the immunoglobulin lambda joining 1 and the immunoglobulin lambda constant 1 gene segments. Alternative splicing results in multiple transcript variants. [provided by RefSeq, May 2010].Specifications
IGLL5 | |
Polyclonal | |
Unconjugated | |
IGLL5 | |
anti HBs antibody light-chain Fab fragment; G lambda-1; Germline immunoglobulin lambda 1; IGL; IGLL5; IGLV; immunoglobin anti-granzymeB light chain variable region; immunoglobulin lambda like polypeptide 5; immunoglobulin lambda-3 surrogate light chain; immunoglobulin lambda-like polypeptide 5; omega light chain protein 14.1; VL-MAR | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100423062 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
B9A064 | |
IGLL5 | |
KLH conjugated synthetic peptide between 167-193 amino acids from the C-terminal region of human IGLL5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |