Learn More
IgM[b] Mouse anti-Mouse, Brilliant Violet 711, Clone: AF6-78, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742347
Additional Details : Weight : 0.01000kg
Description
The AF6-78 antibody reacts specifically with mouse IgM of Igh-C[b] /and related haplotypes (e.g., C57BL/6, C57BL/10, SJL, AKR, NZB). It does not react with IgM of Igh-C[a] /or related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2, PL, RIII). Cross-reaction with IgM of Igh-C[e] /haplotype (e.g., A/J) has been observed. AF6-78 antibody does not react with free μ heavy chain in vitro or in the cytoplasm of pre-B lymphocytes, which lack Ig light chain. It has not been shown to stimulate B-cell proliferation.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.Specifications
IgM[b] | |
AF6-78 | |
Brilliant Violet 711 | |
Igh-6b | |
C57BL/10 mouse splenocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |