missing translation for 'onlineSavingsMsg'
Learn More

IgM, μ Chain Specific Goat anti-Mouse, HRP, Secondary Antibody, Proteintech

Goat anti-Mouse Affinipure Secondary Antibody

£40.00 - £149.00

Specifications

Antigen IgM, μ Chain Specific
Applications Western Blot
Conjugate HRP
Form Liquid
Host Species Goat
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17690387
View Documents
Proteintech
SA00012-6-200UL
200 μL
£40.00
200µL
Please to purchase this item. Need a web account? Register with us today!
17680387
View Documents
Proteintech
SA00012-6-1ML
1 mL
£149.00
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

IgM, μ Chain Specific
HRP
Goat
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Western Blot
Liquid
Mouse
Secondary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications