Learn More
Invitrogen™ IgM Human Uncoated ELISA Kit
For quantitative detection of human IgM.
£368.00 - £558.00
Specifications
Target Species | Human |
---|---|
For Use With (Application) | ELISA |
Synonym | Immunoglobulin M |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15580957
|
Invitrogen™
88-50620-77 |
20 x 96 Tests |
£558.00
Pack of 20 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15500967
|
Invitrogen™
88-50620-88 |
10 x 96 Tests |
£368.00
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
The Human Immunoglobulin M (IgM) Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgM. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Specifications
Human | |
ELISA | |
Immunoglobulin M |
For Research Use Only