Learn More
IgM Mouse Anti-Rat, Brilliant Violet 421, Clone: G53-238, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742499
Additional Details : Weight : 0.01000kg
Description
The G53-238 antibody reacts specifically with rat IgM monomers and pentamers. It does not react with other Ig isotypes. G53-238 antibody has not been shown to stimulate B-cell proliferation.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.Specifications
IgM | |
G53-238 | |
Brilliant Violet 421 | |
Igh-6, Igh-1a, mu-immunoglobulin, RGD1359202 | |
Pooled Rat Ig | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |