Learn More
Mouse IgM Isotype Control (11E10), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Isotype Control
£75.50 - £190.00
Specifications
Antigen | Mouse IgM |
---|---|
Clone | 11E10 |
Conjugate | eFluor 660 |
Form | Liquid |
Host Species | Mouse |
Description
Description: The mouse IgM is useful as an isotype control immunoglobulin. The immunoglobulin was generated by immunization with lipopolysaccharide (LPS). The immunoglobulin is reactive with LPS. Applications Reported: eFluor® 660 Mouse IgM Isotype Control has been reported for use in flow cytometric analysis and immunohistochemical staining. Applications Tested: eFluor® 660 Mouse IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at the same concentration as the experimental antibody. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Specifications
Mouse IgM | |
eFluor 660 | |
Mouse | |
Isotype Control (Primary) |
11E10 | |
Liquid | |
RUO | |
Affinity chromatography |
For Research Use Only.