Learn More
Honeywell Safety Products™ Ignite Glasses
Lightweight, frameless, distortion free, unobstructed vision safety glasses. Honeywell Safety Products™ Ignite Safety Glasses offer a comfortable and secure fit.
Brand: Honeywell Safety Products™ 1017085
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Nine-base, wraparound lens design.
- Close-to-the-face protection and superior coverage.
- Lightweight frameless design.
- All-day comfort and180° of distortion free, unobstructed vision.
- Adjustable nose-piece.
- Reduced slippage and personalized, comfortable, secure fit.
Specifications
Impact-resistant, Hardcoat | |
Safety glasses | |
Black, Silver | |
Polycarbonate |
EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775 | |
Indoor use. Automotive, Building, Construction, Maintenance, Paper, Ship Building, Transportation and Utility industries. | |
1 Ea. | |
Polyamide |