missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA559417
This item is not returnable.
View return policy
Description
Immunogen sequence: KSNPLRNKEI KEDSQKKKSA RRIFQEIAQE LYHKRHVETN QQSEKDNNTY ENRRVLSNYE RP Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 69%.
IGSF6 (DORA), a novel member of the immunoglobulin superfamily (IGSF) from human and rat expressed in dendritic and myeloid cells. Using a probe from the open reading frame of the rat cDNA, we isolated a cosmid which contains the entire mouse gene. By comparative analysis and reverse transcriptase polymerase chain reaction, we defined the intron/exon structure and the mRNA of the mouse gene and, with respect to human BAC clones, the human gene. The genes span 10 kb (mouse) and 12 kb (human), with six exons arranged in a manner similar to other members of the IGSF. All intron/exon boundaries follow the GT-AG rule. Expression of the mouse Igsf6 gene is restricted to cells of the immune system, particularly macrophages. Northern blot revealed a single mRNA of 2.5 kb, in contrast to the human gene which is expressed as two mRNAs of 1 and 2.5 kb. The human and mouse genes were localized to a locus associated with inflammatory bowel disease. Analysis of the flanking regions of the Igsf6 gene revealed the presence of an unrelated gene, transcribed from the opposite strand of the DNA and oriented such that the Igsf6 gene is encoded entirely within an intron. An identical organization is seen in human.Specifications
IGSF6 | |
Polyclonal | |
Unconjugated | |
IGSF6 | |
DORA; down-regulated by activation (immunoglobulin superfamily); IGSF6; Immunoglobulin superfamily member 6; immunoglobulin superfamily, member 6; Protein DORA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10261 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95976 | |
IGSF6 | |
Recombinant protein corresponding to Human IGSF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only