missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgY++ (IgG)(H+L) Donkey anti-Chicken, HRP, Secondary Antibody, Proteintech
Donkey anti-Chicken Affinipure Secondary Antibody
£40.00 - £149.00
Specifications
Antigen | IgY++ (IgG)(H+L) |
---|---|
Applications | ELISA, Western Blot |
Conjugate | HRP |
Form | Liquid |
Host Species | Donkey |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17640297
|
Proteintech
SA00001-14-200UL |
200 μL |
£40.00
200µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17630297
|
Proteintech
SA00001-14-1ML |
1 mL |
£149.00
1mL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IgY++ (IgG)(H+L) | |
HRP | |
Donkey | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
ELISA, Western Blot | |
Liquid | |
Chicken | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |