missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-10 Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-K21
Additional Details : Weight : 0.03500kg
Description
Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.Specifications
P22301 | |
Lot-specific | |
ELISA | |
3586 | |
This Human IL-10 ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hr 45 min |
Lot-specific | |
Biotin | |
Colorimetric Microplate Reader | |
IL10 | |
HRP | |
RUO | |
Human | |
24 hr 30 min |