missing translation for 'onlineSavingsMsg'
Learn More

IL-15 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

£165.00 - £2745.00

Specifications

Antigen IL-15
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17874513
View Documents
Invitrogen
500-P15BT-25UG
25 μg
£165.00
25µg
Please to purchase this item. Need a web account? Register with us today!
17864513
View Documents
Invitrogen
500-P15BT-50UG
50 μg
£212.00
50µg
Please to purchase this item. Need a web account? Register with us today!
17854443
View Documents
Invitrogen
500-P15BT-1MG
2 x 500μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
30054681
View Documents
Invitrogen
500-P15BT-500UG
500μg
£1373.00
500µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: MNWVNVISDL KKIEDLIQSM HIDATLYTES DVHPSCKVTA MKCFLLELQV ISLESGDASI HDTVENLIIL ANNSLSSNGN VTESGCKECE ELEEKNIKEF LQSFVHIVQM FINTS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-15. Anti-Human IL-15-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIL-15 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-15 (500-P15) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIL-15. Western Blot: To detect hIL-15 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIL-15 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P15BT-1MG will be provided as 2 x 500 μg

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
Specifications

Specifications

IL-15
ELISA, Western Blot
Biotin
Rabbit
Human
P40933
3600
E.coli-derived Recombinant Human IL-15
Antigen affinity chromatography
IL15
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
IL15
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications