missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-17F Human ProQuantum Immunoassay Kit
Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument
£648.00 - £2895.00
Specifications
Accession Number | Q96PD4 |
---|---|
Assay Range | 0.0128 to 5000 pg/mL |
Assay Sensitivity | <0.01 pg/mL |
Detection Method | Taqman™ Real-time PCR |
Immunoassay Kit Format | Proximity-based Immunoassay |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16326542
|
Invitrogen™
A35612 |
96 Tests |
£648.00
Pack of 96 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
17173119
|
Invitrogen™
A356125 |
5 x 96 Tests |
£2895.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
Description
- High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
- Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
- Fast, easy workflow: No wash steps, 2 hours from sample to answer
- Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
- No proprietary instrument to purchase: Runs on any real-time PCR instrument
- ProQuantum software available: Robust data analysis and statistical group wise comparison
Specifications
Q96PD4 | |
<0.01 pg/mL | |
Proximity-based Immunoassay | |
Human | |
qPCR Instruments | |
1 hr. | |
FAM™ | |
Serum,2-5μL; Plasma,2-5μL; Supernatant,2-5μL | |
Human | |
2 hr. |
0.0128 to 5000 pg/mL | |
Taqman™ Real-time PCR | |
ProQuantum | |
Protein quantitation | |
112744 | |
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard | |
RUO | |
Frozen at -20°C | |
30 min. |