missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-20 Mouse ELISA Kit

Sandwich ELISA Kit

£504.00 - £2310.00

Specifications

Accession Number Q9JKV9
Assay Sensitivity 16 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485293
View Documents
Invitrogen™
EMIL20
96 Tests
£504.00
1 set
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
17155749
View Documents
Invitrogen™
EMIL20X5
5 x 96 Tests
£2310.00
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

The Mouse Interleukin-20 (Ms IL-20) ELISA quantitates Ms IL-20 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-20. Principle of the method The Mouse IL-20 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Cytokines that signal through Class II receptors form a distinct family that includes the interferons and interleukin 10 (IL-10). Recent identification of several IL-10 homologs has defined a cytokine subfamily that includes AK155, IL-19, IL-20, IL-22, and IL-24. IL-20 belongs to the IL-10 family and plays a role in skin inflammation and the development of hematopoietic cells. Little is known about its other biological functions and clinical implications. These cytokines display pro-inflammatory activities in different tissues, including skin.
Specifications

Specifications

Q9JKV9
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
58181
<12%
HRP
Oncology
Zcyto10
1 hr. 20 min.
16 pg/mL
Biotin
Cytokines & Receptors
Mouse
Colorimetric Microplate Reader
ZCYTO10
<10%
RUO
2°C to 8°C
Mouse
4 hr. 45 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.