missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-27 R alpha/WSX-1/TCCR Antibody, R&D Systems™
Goat Polyclonal Antibody
£147.94 - £393.97
Specifications
Antigen | IL-27R alpha/WSX-1/TCCR |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749027
|
R&D Systems
AF2109-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18000322
|
R&D Systems
AF2109 |
100 μg |
£393.97
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-27R alpha/WSX-1/TCCR Polyclonal specifically detects IL-27R alpha/WSX-1/TCCR in Mouse samples. It is validated for Western Blot.Specifications
IL-27R alpha/WSX-1/TCCR | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
IL27RA | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O70394 | |
9466 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-27 R alpha/WSX-1/TCCR Gly29-Lys510 Accession # O70394 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-27 R alpha/WSX-1/TCCR in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant human TCCR is observed. |
For Research Use Only