missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

£177.00 - £2745.00

Specifications

Antigen IL-3
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17885133
View Documents
PeproTech
500-P53BT-25UG
25 μg
£177.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17855053
View Documents
PeproTech
500-P53BT-50UG
50 μg
£228.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17855433
View Documents
Invitrogen
500-P53BT-1MG
2 x 500μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30054575
View Documents
PeproTech
500-P53BT-500UG
500 μg
£1466.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-3. Anti-Murine IL-3-specific antibody was purified by affinity chromatography and then biotinylated. Lyophilized from sterile-filtered PBS solution. Sandwich ELISA: To detect mIL-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-3 (500-P53) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mIL-3. Western Blot: To detect mIL-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P53BT-1MG will be provided as 2 x 500 μg

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications

Specifications

IL-3
ELISA, Western Blot
Biotin
Rabbit
Mouse
P01586
16187
E.coli-derived Recombinant Murine IL-3.
Antigen affinity chromatography
IL3
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF
IL3
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.