missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-9 Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-K20
Additional Details : Weight : 0.03500kg
Description
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.Specifications
P15248 | |
Lot-specific | |
ELISA | |
3578 | |
This Human IL-9 ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hr 45 min |
Lot-specific | |
Biotin | |
Colorimetric Microplate Reader | |
Il9 | |
HRP | |
RUO | |
Human | |
24 hr 30 min |