missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL13 Recombinant Rat Protein
Recombinant IL13 is a bioactive protein intended for use in cell culture applications.
Brand: Gibco™ PRC0131
Code : 50
Additional Details : Weight : 0.50000kg
Description
IL13 down-regulates macrophage produced cytokines such as TNF-&alpha, IL-1, IL-6, IL-8, and IL-12 in response to LPS challenge or IFN-γ. IL13 induces monocyte differentiation and also induces B cell differentiation and proliferation.Specifications
Cell Culture | |
12.7 kDa | |
100μg | |
< 0.1ng/μg | |
Rat | |
Cytokines & Receptors | |
Interleukins |
116553 | |
Sequential Chromatography | |
Store in refrigerator (2° to 8°C). | |
IL-13 | |
E. coli | |
Lyophilized | |
95% |
For Research Use Only. Not for use in diagnostic procedures.