missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL20 Recombinant Mouse Protein
Recombinant IL20 is a bioactive protein intended for use in cell culture applications.
Brand: Gibco™ PMC0201
Code : 50
Additional Details : Weight : 0.50000kg
Description
Recombinant IL20 is a bioactive protein intended for use in cell culture applications. There are two heterodimeric receptor complexes for IL20. The first is composed of IL20 Ra and IL20 Rb. The second is composed of IL-22 R and IL20b. IL20 induces the proliferation of multipotential hematopoietic progenitor cells, directs the differentiation and expansion of keratinocytes, and promotes the release of proinflammatory mediators in keratinocytes and other cells expressing the IL20 receptor.Specifications
Cell Culture | |
Sequential Chromatography | |
Store in refrigerator (2–8°C) | |
IL-20 | |
Recombinant | |
Cytokines & Receptors | |
Interleukins |
∼18 kDa | |
100μg | |
< 0.1 ng/μg | |
Mouse | |
E. coli | |
Lyophilized | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.