missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL22RA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13462-1-AP-150UL
This item is not returnable.
View return policy
Description
A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.Specifications
IL22RA1 | |
Polyclonal | |
Unconjugated | |
IL22RA1 | |
CRF2 9, IL 22 receptor subunit alpha 1, IL 22R alpha 1, IL 22RA1, IL22R, IL22R1, IL22RA1, ZcytoR11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230828, 362629, 58985 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80XZ4, Q8N6P7 | |
Il22ra1 | |
IL22RA1 Fusion Protein Ag4056 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |