missing translation for 'onlineSavingsMsg'
Learn More

IL2RB Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Brand:  Abnova PAB30302.100uL

Product Code. 16187700

  • £426.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rabbit polyclonal antibody raised against partial recombinant human IL2RB.

  • High specificity
  • High reactivity
Specifications

Specifications

IL2RB
Polyclonal
Rabbit polyclonal antibody raised against partial recombinant human IL2RB.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
CD122/P70-75
Rabbit
Antigen affinity purification
RUO
3560
Store at 4°C for short term storage. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000 - 1:2500) The optimal working dilution should be determined by the end user.
P14784
IL2RB
Recombinant protein corresponding to amino acids 66-122 of human IL2RB.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)